Assay Details
Target Gene Details
Entrez Gene ID: | 230025 |
Gene Name: | PR domain containing 13 |
Gene Aliases: |
- |
Location: |
Chr.4:21675865-21685963 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prdm13 | NM_001080771.1 | 4 | 2358 | NP_001074240.1 |
More Information
Set Membership: |
|

Genomic Map