Assay Details
Target Gene Details
Entrez Gene ID: | 71951 |
Gene Name: | glypican 2 (cerebroglycan) |
Gene Aliases: |
2410016G05Rik |
Location: |
Chr.5:138273657-138279958 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpc2 | NM_172412.2 | 3 | 668 | NP_766000.1 |
| AK010515.1 | 3 | 698 | ||
| AK048605.1 | 3 | 611 | ||
| AK049639.1 | 3 | 669 | BAC33852.1 | |
| AK160959.1 | 3 | 714 | BAE36115.1 | |
| BC083180.1 | 3 | 675 | AAH83180.1 |
Target Gene Details
Entrez Gene ID: | 50878 |
Gene Name: | stromal antigen 3 |
Gene Aliases: |
SA-2 |
Location: |
Chr.5:138272589-138312397 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stag3 |

Genomic Map