Assay Details
Target Gene Details
Entrez Gene ID: | 230459 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 13 |
Gene Aliases: |
- |
Location: |
Chr.4:96042660-96077566 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j13 | NM_145548.4 | 9 | 1904 | NP_663523.2 |
| AK143903.1 | 9 | 1905 | BAE25592.1 | |
| AK143981.1 | BAE25643.1 | |||
| AK165405.1 | 9 | 1904 | BAE38165.1 | |
| AK165473.1 | 9 | 1882 | BAE38210.1 |
More Information
Set Membership: |
|

Genomic Map