Assay Details
Target Gene Details
Entrez Gene ID: | 74519 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 9 |
Gene Aliases: |
8430417E17Rik |
Location: |
Chr.4:96568429-96591719 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j9 | NM_028979.2 | 1 | 291 | NP_083255.1 |
| AF336850.1 | 1 | 291 | AAK59868.1 | |
| AK018422.1 | 1 | 235 | BAB31202.1 | |
| AK033961.1 | 1 | 261 | ||
| AK037031.1 | 1 | 387 | ||
| BC119556.2 | 1 | 215 |
More Information
Set Membership: |
|

Genomic Map