Assay Details
Target Gene Details
Entrez Gene ID: | 230082 |
Gene Name: | nucleolar protein family 6 (RNA-associated) |
Gene Aliases: |
AA410091, Nrap |
Location: |
Chr.4:41114427-41124339 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol6 | NM_139236.3 | 26 | 3759 | NP_631982.2 |
| AF361077.1 | 26 | 3708 | AAL74401.1 | |
| AF361078.1 | AAL74402.1 | |||
| AK089138.1 | ||||
| AK155818.1 | 26 | 3698 | BAE33447.1 | |
| AK166559.1 | 26 | 3686 | ||
| AK166650.1 | 26 | 3685 | BAE38917.1 | |
| AK190299.1 | 1 | 161 | ||
| AK192663.1 | 1 | 161 | ||
| AK193940.1 | 1 | 161 | ||
| AK194986.1 | 1 | 161 | ||
| AK197279.1 | 1 | 161 | ||
| AK202606.1 | 1 | 161 | ||
| AK203818.1 | 1 | 161 | ||
| BC019981.1 | 12 | 1714 | AAH19981.1 | |
| BC021856.1 | 12 | 1764 | AAH21856.1 | |
| BC059820.1 | 26 | 3677 |
More Information
Set Membership: |
|

Genomic Map