Assay Details
Target Gene Details
Entrez Gene ID: | 66625 |
Gene Name: | PNN interacting serine/arginine-rich |
Gene Aliases: |
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18 |
Location: |
Chr.4:21847583-21876475 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnisr | NM_025669.1 | 12 | 3497 | NP_079945.1 |
| AK031964.1 | 12 | 3034 | ||
| AK035634.1 | 1 | 1134 | BAC29132.1 | |
| BC141031.1 | 12 | 3361 |

Genomic Map