Assay Details
Target Gene Details
Entrez Gene ID: | 69876 |
Gene Name: | THAP domain containing, apoptosis associated protein 3 |
Gene Aliases: |
2010013E08Rik, 2210418H06Rik |
Location: |
Chr.4:151982638-151988986 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thap3 | NM_001145929.1 | 3 | 421 | NP_001139401.1 |
| NM_175152.4 | 4 | 523 | NP_780361.2 | |
| AK008225.1 | 4 | 495 | ||
| AK038184.1 | 4 | 497 | BAC29945.1 | |
| AK050740.1 | 4 | 537 | BAC34401.1 | |
| BC094395.1 | 4 | 504 | AAH94395.1 |

Genomic Map