Assay Details
Target Gene Details
Entrez Gene ID: | 21885 |
Gene Name: | transducin-like enhancer of split 1, homolog of Drosophila E(spl) |
Gene Aliases: |
C230057C06Rik, Estm14, Grg-1, Grg1, Tle4l |
Location: |
Chr.4:72109945-72200893 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tle1 | NM_001285529.1 | 1 | 287 | NP_001272458.1 |
| NM_001285530.1 | 1 | 287 | NP_001272459.1 | |
| NM_001285531.1 | 1 | 287 | NP_001272460.1 | |
| NM_001285532.1 | 1 | 287 | NP_001272461.1 | |
| NM_011599.5 | 1 | 287 | NP_035729.3 | |
| AK044642.1 | 1 | 288 | ||
| AK046402.1 | 1 | 254 | BAC32708.1 | |
| AK048515.1 | 1 | 249 | ||
| AK082499.1 | 1 | 252 | BAC38509.1 | |
| AY155196.1 | 1 | 256 | AAN77515.1 | |
| BB619055.1 | 1 | 313 | ||
| BC057573.1 | 1 | 117 | AAH57573.1 |
More Information
Set Membership: |
|

Genomic Map