Assay Details
Target Gene Details
Entrez Gene ID: | 21885 |
Gene Name: | transducin-like enhancer of split 1, homolog of Drosophila E(spl) |
Gene Aliases: |
C230057C06Rik, Estm14, Grg-1, Grg1, Tle4l |
Location: |
Chr.4:72109945-72200893 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tle1 | NM_001285529.1 | 20 | 3722 | NP_001272458.1 |
| NM_001285530.1 | 18 | 3503 | NP_001272459.1 | |
| NM_011599.5 | 20 | 3725 | NP_035729.3 | |
| AK082499.1 | 20 | 3691 | BAC38509.1 |
More Information
Set Membership: |
|

Genomic Map