Assay Details
Target Gene Details
Entrez Gene ID: | 243270 |
Gene Name: | hydrocarboxylic acid receptor 1 |
Gene Aliases: |
Gpr81 |
Location: |
Chr.5:123876736-123880020 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcar1 | NM_175520.4 | 1 | 2035 | NP_780729.2 |
| AK029064.1 | 1 | 2020 | BAC26273.1 | |
| AK046700.1 | 1 | 2036 |

Genomic Map