Assay Details
Target Gene Details
Entrez Gene ID: | 67389 |
Gene Name: | family with sequence similarity 132, member A |
Gene Aliases: |
1110035L05Rik, C1qdc2, CTRP12, alipolin |
Location: |
Chr.4:155962312-155966629 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam132a | NM_026125.3 | 1 | 81 | NP_080401.1 |
| AK002876.1 | 1 | 63 | BAB22423.1 | |
| AK004116.1 | 1 | 76 | BAB23178.1 | |
| AK007335.1 | 1 | 68 | ||
| BC026939.1 | AAH26939.1 |
More Information
Set Membership: |
|

Genomic Map