Assay Details
Target Gene Details
Entrez Gene ID: | 75811 |
Gene Name: | stablizer of axonemal microtubules 1 |
Gene Aliases: |
4930500O09Rik, Fam154a |
Location: |
Chr.4:86444698-86558303 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Saxo1 | NM_001081096.1 | 4 | 1099 | NP_001074565.1 |
| AK015673.1 | 4 | 1071 | ||
| AK049262.1 | 4 | 923 |

Genomic Map