Assay Details
Target Gene Details
Entrez Gene ID: | 329831 |
Gene Name: | family with sequence similarity 166, member B |
Gene Aliases: |
4833436C18Rik |
Location: |
Chr.4:43426675-43431593 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam166b | NM_001162381.1 | 5 | 1161 | NP_001155853.1 |
| NM_177377.5 | 4 | 1238 | NP_796351.2 | |
| AK029435.1 | 4 | 1239 | BAC26451.1 | |
| AK053208.1 | 4 | 1239 | BAC35311.1 |
Target Gene Details
Entrez Gene ID: | 100213 |
Gene Name: | RUN and SH3 domain containing 2 |
Gene Aliases: |
AI840675, mKIAA0375 |
Location: |
Chr.4:43381982-43427092 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rusc2 | NM_001037709.2 | NP_001032798.1 | ||
| NM_199057.3 | NP_951012.2 | |||
| AI849772.1 | ||||
| AK122263.1 | BAC65545.1 | |||
| AK132170.1 | BAE21009.1 | |||
| AK166260.1 | ||||
| BC024790.1 | AAH24790.1 |

Genomic Map