Assay Details
Target Gene Details
Entrez Gene ID: | 23957 |
Gene Name: | nuclear receptor subfamily 0, group B, member 2 |
Gene Aliases: |
SHP, SHP-1, Shp1 |
Location: |
Chr.4:133553390-133556536 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr0b2 | NM_011850.2 | 2 | 1032 | NP_035980.1 |
| AK004930.1 | 2 | 1042 | ||
| BC019540.1 | 2 | 1032 | AAH19540.1 |
More Information
Set Membership: |
|

Genomic Map