Assay Details
Target Gene Details
Entrez Gene ID: | 625638 |
Gene Name: | family with sequence similarity 43, member B |
Gene Aliases: |
OTTMUSG00000009974 |
Location: |
Chr.4:138394092-138396458 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam43b | NM_001081672.2 | 1 | 787 | NP_001075141.1 |
| AK134570.1 | 1 | 787 |
More Information
Set Membership: |
|

Genomic Map