Assay Details
Target Gene Details
Entrez Gene ID: | 68099 |
Gene Name: | family with sequence similarity 92, member A |
Gene Aliases: |
6720467C03Rik, Fam92a1 |
Location: |
Chr.4:12153722-12172022 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam92a | NM_026558.4 | 1 | 111 | NP_080834.3 |
| AK032690.1 | 1 | 91 | ||
| AK078438.1 | 1 | 92 | BAC37273.1 | |
| AK088705.1 | 1 | 94 | BAC40516.1 | |
| CJ064072.1 | 1 | 112 |

Genomic Map