Assay Details
Target Gene Details
Entrez Gene ID: | 17096 |
Gene Name: | Yamaguchi sarcoma viral (v-yes-1) oncogene homolog |
Gene Aliases: |
AA407514, Hck-2, p53Lyn, p56Lyn |
Location: |
Chr.4:3678121-3791612 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lyn | NM_001111096.1 | 13 | 2520 | NP_001104566.1 |
| NM_010747.2 | 13 | 2457 | NP_034877.2 | |
| AK028112.1 | 13 | 2453 | BAC25753.1 | |
| AK151914.1 | 13 | 2516 | ||
| AK152199.1 | 13 | 2518 | BAE31028.1 | |
| AK153038.1 | 13 | 2454 | BAE31669.1 | |
| AK170561.1 | 13 | 2518 | BAE41882.1 | |
| BC003253.1 | 13 | 2397 | ||
| BC031547.1 | 13 | 2490 | AAH31547.1 | |
| BC157998.1 | 13 | 2289 | ||
| M57696.1 | 13 | 2356 | AAA39471.1 | |
| M57697.1 | 13 | 2293 | AAA39472.1 |

Genomic Map