Assay Details
Target Gene Details
Entrez Gene ID: | 242915 |
Gene Name: | GRB2 associated, regulator of MAPK1-like |
Gene Aliases: |
Fam59b, Gm444 |
Location: |
Chr.5:30105196-30118380 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gareml | NM_001167879.1 | 6 | 2618 | NP_001161351.1 |
| BC060268.1 | 5 | 2402 | AAH60268.1 | |
| BC113782.1 | 4 | 2456 | AAI13783.1 | |
| BC113783.1 | 4 | 2457 |

Genomic Map