Assay Details
Target Gene Details
Entrez Gene ID: | 329877 |
Gene Name: | DENN/MADD domain containing 4C |
Gene Aliases: |
1700065A05Rik, AA420392 |
Location: |
Chr.4:86748555-86850603 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd4c | NM_184088.1 | 2 | 412 | NP_908976.1 |
| AK155220.1 | 2 | 413 | BAE33131.1 | |
| BC146602.1 |

Genomic Map