Assay Details
Target Gene Details
Entrez Gene ID: | 68777 |
Gene Name: | transmembrane protein 53 |
Gene Aliases: |
1110038M16Rik, 1500001P22Rik, 1700012A05Rik |
Location: |
Chr.4:117251951-117268588 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem53 | NM_001285812.1 | 3 | 439 | NP_001272741.1 |
| NM_001285814.1 | 4 | 504 | NP_001272743.1 | |
| NM_001285815.1 | 4 | 522 | NP_001272744.1 | |
| NM_001285816.1 | 2 | 302 | NP_001272745.1 | |
| NM_026837.3 | 3 | 418 | NP_081113.1 | |
| NM_029589.1 | 4 | 340 | NP_083865.1 | |
| AK004164.1 | 3 | 381 | BAB23200.1 | |
| AK005104.1 | 3 | 325 | ||
| AK005885.1 | 4 | 344 | BAB24300.1 | |
| BC019937.1 | 3 | 344 | AAH19937.1 | |
| BC020121.1 | 3 | 348 | AAH20121.1 |

Genomic Map