Assay Details
Target Gene Details
Entrez Gene ID: | 13347 |
Gene Name: | DNA fragmentation factor, alpha subunit |
Gene Aliases: |
A330085O09Rik, DFF35, Dff45, ICAD, ICAD-L, ICAD-S |
Location: |
Chr.4:149104142-149120653 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dffa | NM_001025296.2 | 6 | 1681 | NP_001020467.1 |
| NR_104263.1 | 4 | 1115 | ||
| AK051011.1 | 6 | 1611 | BAC34496.1 | |
| AK154488.1 | 6 | 1645 |

Genomic Map