Assay Details
Target Gene Details
Entrez Gene ID: | 17357 |
Gene Name: | MARCKS-like 1 |
Gene Aliases: |
AL022768, AW215397, AW536807, D4Bc1, F52, MacMARCKS, Macs2, Macs3, Mlp, Mrp |
Location: |
Chr.4:129513581-129515981 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Marcksl1 | NM_010807.4 | 1 | 241 | NP_034937.1 |
| AK079390.1 | 1 | 190 | BAC37630.1 | |
| AK083913.1 | 1 | 192 | BAC39058.1 | |
| AK150261.1 | 1 | 189 | ||
| AK152990.1 | 1 | 189 | BAE31636.1 | |
| AK169355.1 | 1 | 192 | BAE41104.1 | |
| BC006757.1 | 1 | 169 | AAH06757.1 | |
| CX243729.1 | 1 | 241 | ||
| X61399.1 | 1 | 178 | CAA43671.1 |

Genomic Map