Assay Details
Target Gene Details
Entrez Gene ID: | 50793 |
Gene Name: | origin recognition complex, subunit 3 |
Gene Aliases: |
Orc3l |
Location: |
Chr.4:34566781-34614942 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orc3 | NM_001159563.1 | 20 | 3482 | NP_001153035.1 |
| NM_015824.4 | 20 | 3485 | NP_056639.3 | |
| AK041216.1 | 1 | 1067 | ||
| AK086579.1 | 20 | 3488 | BAC39695.1 | |
| AK163854.1 | 1 | 1020 |
More Information
Set Membership: |
|

Genomic Map