Assay Details
Target Gene Details
Entrez Gene ID: | 75172 |
Gene Name: | coiled-coil domain containing 146 |
Gene Aliases: |
4930528G09Rik, mKIAA1505 |
Location: |
Chr.5:21292961-21424677 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc146 | NM_029195.1 | NP_083471.1 | ||
| AK015926.1 | BAB30036.1 |
Target Gene Details
Entrez Gene ID: | 14190 |
Gene Name: | fibrinogen-like protein 2 |
Gene Aliases: |
AI385601, musfiblp |
Location: |
Chr.5:21372673-21378386 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgl2 | NM_008013.4 | 1 | 422 | NP_032039.2 |
| AK009500.1 | 1 | 454 | ||
| AK036402.1 | 1 | 393 | BAC29413.1 | |
| AK036784.1 | 1 | 389 | ||
| BC028893.1 | 1 | 422 | AAH28893.1 |
More Information
Set Membership: |
|

Genomic Map