Assay Details
Target Gene Details
Entrez Gene ID: | 231630 |
Gene Name: | FIC domain containing |
Gene Aliases: |
D5Ertd40e, Hype |
Location: |
Chr.5:113735677-113740607 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ficd | NM_001010825.3 | 3 | 1046 | NP_001010825.2 |
| AK044446.1 | 3 | 855 | BAC31924.1 | |
| AK054325.1 | 3 | 811 | BAC35731.1 | |
| AK087477.1 | 3 | 1030 | BAC39889.1 | |
| BC139821.1 | 2 | 683 |

Genomic Map