Assay Details
Target Gene Details
Entrez Gene ID: | 140477 |
Gene Name: | diencephalon/mesencephalon homeobox 1 |
Gene Aliases: |
Atx, Cdmx, Mbx, Otx3 |
Location: |
Chr.4:115915119-115939926 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmbx1 | NM_001025567.1 | 4 | 555 | NP_001020738.1 |
| NM_130865.3 | 4 | 570 | NP_570935.3 | |
| AB037698.1 | 4 | 536 | BAC00919.1 | |
| AF421857.1 | 3 | 516 | AAL30508.1 | |
| AF421858.1 | 3 | 501 | AAL30509.1 | |
| AF448118.1 | 4 | 554 | AAN76724.1 | |
| AF461039.1 | 3 | 519 | AAL66343.1 | |
| AF463513.1 | 3 | 516 | AAL68836.1 | |
| AF499446.1 | 3 | 2337 | AAM78514.1 | |
| AK141654.1 | 4 | 571 | ||
| BC050912.1 | 4 | 532 | AAH50912.1 | |
| BY732319.1 | 4 | 555 | ||
| CN455087.1 | 3 | 472 |
More Information
Set Membership: |
|

Genomic Map