Assay Details
Target Gene Details
Entrez Gene ID: | 140477 |
Gene Name: | diencephalon/mesencephalon homeobox 1 |
Gene Aliases: |
Atx, Cdmx, Mbx, Otx3 |
Location: |
Chr.4:115915119-115939926 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmbx1 | NM_001025567.1 | 5 | 3470 | NP_001020738.1 |
| NM_130865.3 | 5 | 3485 | NP_570935.3 | |
| AF499446.1 | 4 | 5252 | AAM78514.1 | |
| AK141654.1 | 5 | 3486 | ||
| BC050912.1 | 5 | 3447 | AAH50912.1 |
More Information
Set Membership: |
|

Genomic Map