Assay Details
Target Gene Details
Entrez Gene ID: | 242466 |
Gene Name: | zinc finger protein 462 |
Gene Aliases: |
6030417H05, 9430078C22Rik, Gt4-2, Zfpip |
Location: |
Chr.4:54944583-55083563 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp462 | NM_172867.3 | 8 | 6798 | NP_766455.2 |
| AK031378.1 | 3 | 309 | BAC27374.1 | |
| AK173266.1 | 6 | 1111 | BAD32544.1 | |
| BC099692.1 | 3 | 395 | AAH99692.1 | |
| DQ355518.1 | 7 | 6495 | ||
| X66902.1 | 4 | 501 | CAA47353.1 |
More Information
Set Membership: |
|

Genomic Map