Assay Details
Target Gene Details
Entrez Gene ID: | 22704 |
Gene Name: | zinc finger protein 46 |
Gene Aliases: |
AI426962, Mlz-4, Zfp-46, Znf436 |
Location: |
Chr.4:136284168-136293942 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp46 | NM_009557.3 | 1 | 132 | NP_033583.2 |
| AK051031.1 | 1 | 105 | BAC34503.1 | |
| AK053652.1 | 1 | 135 | BAC35464.1 | |
| AK136405.1 | BAE22966.1 | |||
| AK148255.1 | 1 | 81 | BAE28441.1 | |
| AK173240.1 | 1 | 119 | BAD32518.1 | |
| BB665308.1 | 1 | 132 |
More Information
Set Membership: |
|

Genomic Map