Assay Details
Target Gene Details
Entrez Gene ID: | 22704 |
Gene Name: | zinc finger protein 46 |
Gene Aliases: |
AI426962, Mlz-4, Zfp-46, Znf436 |
Location: |
Chr.4:136284168-136293942 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp46 | NM_009557.3 | 3 | 3310 | NP_033583.2 |
| AK136405.1 | 3 | 3219 | BAE22966.1 | |
| AK148255.1 | 3 | 3259 | BAE28441.1 | |
| AK173240.1 | 3 | 3297 | BAD32518.1 | |
| BC006587.1 | AAH06587.1 | |||
| BC092398.1 | 1 | 2620 | AAH92398.1 | |
| M98502.1 | 1 | 2833 | AAA39949.1 |
More Information
Set Membership: |
|

Genomic Map