Assay Details
Target Gene Details
Entrez Gene ID: | 11553 |
Gene Name: | adrenergic receptor, alpha 2c |
Gene Aliases: |
Adra-2c, [a]2C, alpha2-C4, alpha2C |
Location: |
Chr.5:35278566-35281763 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra2c | NM_007418.3 | 1 | 2127 | NP_031444.2 |
More Information
Set Membership: |
|

Genomic Map