Assay Details
Target Gene Details
Entrez Gene ID: | 15502 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily A, member 1 |
Gene Aliases: |
Hsj2, Nedd7 |
Location: |
Chr.4:40722468-40737153 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaja1 | NM_001164671.1 | 9 | 2685 | NP_001158143.1 |
| NM_001164672.1 | 9 | 2542 | NP_001158144.1 | |
| NM_008298.5 | 9 | 2636 | NP_032324.1 | |
| AK083046.1 | 9 | 2572 | BAC38744.1 | |
| AK163758.1 | 1 | 337 |
More Information
Set Membership: |
|

Genomic Map