Assay Details
Target Gene Details
Entrez Gene ID: | 56323 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 5 |
Gene Aliases: |
1110058L06Rik, AI462558, Hsc40, Hsp40-3 |
Location: |
Chr.4:42949827-42959425 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb5 | NM_019874.3 | 3 | 1829 | NP_063927.1 |
| AK004300.1 | 1 | 463 | ||
| AK158972.1 | 4 | 2129 | BAE34751.1 | |
| BC057087.1 | 3 | 1829 | AAH57087.1 |
More Information
Set Membership: |
|

Genomic Map