Assay Details
Target Gene Details
Entrez Gene ID: | 15221 |
Gene Name: | forkhead box D3 |
Gene Aliases: |
CWH3, Genesis, Hfh2 |
Location: |
Chr.4:99656299-99658671 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxd3 | NM_010425.3 | 1 | 1979 | NP_034555.3 |
| AF067421.2 | 1 | 1979 | AAC28352.2 | |
| U41047.1 | 1 | 1979 | AAA87569.1 | |
| W87115.1 |
More Information
Set Membership: |
|

Genomic Map