Assay Details
Target Gene Details
Entrez Gene ID: | 194219 |
Gene Name: | schlafen like 1 |
Gene Aliases: |
4933406A14Rik |
Location: |
Chr.4:120531981-120536661 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slfnl1 | NM_177570.3 | 2 | 347 | NP_808238.1 |
| AK077106.1 | 2 | 348 | BAC36615.1 | |
| BC117545.1 | 1 | 98 | ||
| BC118052.1 | 1 | 98 |

Genomic Map