Assay Details
Target Gene Details
Entrez Gene ID: | 194219 |
Gene Name: | schlafen like 1 |
Gene Aliases: |
4933406A14Rik |
Location: |
Chr.4:120531981-120536661 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slfnl1 | NM_177570.3 | 5 | 1537 | NP_808238.1 |
| AK077106.1 | 5 | 1538 | BAC36615.1 | |
| BC117545.1 | 4 | 1285 | ||
| BC118052.1 | 4 | 1288 |

Genomic Map