Assay Details
Target Gene Details
Entrez Gene ID: | 252838 |
Gene Name: | thymocyte selection-associated high mobility group box |
Gene Aliases: |
1700007F02Rik |
Location: |
Chr.4:6686353-6991552 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tox | NM_145711.4 | 1 | 278 | NP_663757.3 |
| AF472514.1 | 1 | 74 | AAL78656.1 | |
| AK029694.1 | 1 | 75 | BAC26568.1 | |
| AK038671.1 | 1 | 279 | BAC30091.1 | |
| AK051947.1 | 2 | 256 | BAC34818.1 | |
| AK133397.1 | 1 | 75 | ||
| AK161578.1 | ||||
| AK161718.1 | ||||
| AK169546.1 | 2 | 144 | ||
| BC080732.1 | 1 | 197 | AAH80732.1 | |
| BY240192.1 | 1 | 355 |

Genomic Map