Assay Details
Target Gene Details
Entrez Gene ID: | 252838 |
Gene Name: | thymocyte selection-associated high mobility group box |
Gene Aliases: |
1700007F02Rik |
Location: |
Chr.4:6686353-6991552 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tox | NM_145711.4 | 7 | 1430 | NP_663757.3 |
| AF472514.1 | 7 | 1226 | AAL78656.1 | |
| AK029694.1 | 7 | 1227 | BAC26568.1 | |
| AK038671.1 | 7 | 1433 | BAC30091.1 | |
| AK133397.1 | 7 | 1227 | ||
| AK161578.1 | 7 | 1216 | ||
| BC080732.1 | 7 | 1347 | AAH80732.1 |

Genomic Map