Assay Details
Target Gene Details
Entrez Gene ID: | 67694 |
Gene Name: | intraflagellar transport 74 |
Gene Aliases: |
1700029H06Rik, Ccdc2, Cmg1, b2b796Clo |
Location: |
Chr.4:94603266-94693233 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift74 | NM_026319.3 | 20 | 2047 | NP_080595.2 |
| AK006492.1 | 11 | 1154 | BAB24617.1 | |
| CD740935.1 | 1 | 111 |

Genomic Map