Assay Details
Target Gene Details
Entrez Gene ID: | 71904 |
Gene Name: | progestin and adipoQ receptor family member VII |
Gene Aliases: |
2310021M12Rik, AV001994, BB007237, Mpra, PGLP, mPR |
Location: |
Chr.4:134496761-134510237 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paqr7 | NM_001285845.1 | 3 | 1806 | NP_001272774.1 |
| NM_001285846.1 | 4 | 1732 | NP_001272775.1 | |
| NM_001285847.1 | 3 | 1695 | NP_001272776.1 | |
| NM_001285849.1 | 3 | 1579 | NP_001272778.1 | |
| NM_027995.3 | 2 | 1611 | NP_082271.1 | |
| AF313618.1 | 2 | 1365 | AAO47231.1 | |
| AK009450.1 | 2 | 1396 | BAB26296.1 | |
| AK078685.1 | 2 | 1369 | ||
| AK161542.1 | 3 | 1453 | BAE36452.1 | |
| AK170307.1 | 4 | 1516 | BAE41704.1 | |
| AY424296.1 | 1 | 1340 | AAR08384.1 | |
| BC022922.1 | 1 | 1340 | AAH22922.1 |
More Information
Set Membership: |
|

Genomic Map