Assay Details
Target Gene Details
Entrez Gene ID: | 230577 |
Gene Name: | prolyl-tRNA synthetase (mitochondrial)(putative) |
Gene Aliases: |
BC027073 |
Location: |
Chr.4:106651021-106655282 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pars2 | NM_001083887.2 | 2 | 2711 | NP_001077356.1 |
| NM_001285783.1 | 2 | 2728 | NP_001272712.1 | |
| NM_001285784.1 | 2 | 1879 | NP_001272713.1 | |
| NM_172272.2 | 2 | 1862 | NP_758476.2 | |
| AK154150.1 | 2 | 2684 | BAE32409.1 | |
| BC027073.1 | 1 | 1552 | AAH27073.1 | |
| BC038817.1 | 2 | 1835 | AAH38817.1 |
More Information
Set Membership: |
|

Genomic Map