Assay Details
Target Gene Details
Entrez Gene ID: | 72397 |
Gene Name: | RNA binding motif protein 12 B1 |
Gene Aliases: |
3000004N20Rik, Rbm12b, Rbm12ba |
Location: |
Chr.4:12138957-12146745 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm12b1 | NM_028226.2 | 1 | 72 | NP_082502.2 |
More Information
Set Membership: |
|

Genomic Map