Assay Details
Target Gene Details
Entrez Gene ID: | 15502 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily A, member 1 |
Gene Aliases: |
Hsj2, Nedd7 |
Location: |
Chr.4:40722468-40737153 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaja1 | NM_001164671.1 | 9 | 5283 | NP_001158143.1 |
| NM_001164672.1 | 9 | 5140 | NP_001158144.1 | |
| NM_008298.5 | 9 | 5234 | NP_032324.1 | |
| AI852330.1 | 1 | 350 | ||
| AK163758.1 | 1 | 2935 |
Target Gene Details
Entrez Gene ID: | 74255 |
Gene Name: | smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans) |
Gene Aliases: |
2600001O03Rik, 2610203K23Rik, AB044414, AI845086, AW556129, Bwd |
Location: |
Chr.4:40735649-40757885 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smu1 | NM_021535.4 | 12 | 2112 | NP_067510.3 |
| AK019174.1 | 12 | 2071 | ||
| AK145982.1 | 12 | 2111 | BAE26804.1 |
More Information
Set Membership: |
|

Genomic Map