Assay Details
Target Gene Details
Entrez Gene ID: | 433766 |
Gene Name: | tripartite motif-containing 63 |
Gene Aliases: |
MuRF1, RF1, Rnf28 |
Location: |
Chr.4:134315088-134329629 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim63 | NM_001039048.2 | 1 | 121 | NP_001034137.2 |
| AK052911.1 | BAC35199.1 | |||
| BC147663.1 | 1 | 89 | ||
| BC147666.1 | 1 | 89 | ||
| CJ131783.1 | 1 | 122 | ||
| CJ131880.1 | 1 | 122 | ||
| DQ229108.1 | ABB16283.1 |
More Information
Set Membership: |
|

Genomic Map