Assay Details
Target Gene Details
Entrez Gene ID: | 231871 |
Gene Name: | diacylglycerol lipase, beta |
Gene Aliases: |
E330036I19Rik |
Location: |
Chr.5:143464487-143504442 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daglb | NM_144915.3 | 1 | 50 | NP_659164.2 |
| AK086747.1 | BAC39734.1 | |||
| BC016105.1 | AAH16105.2 | |||
| BY314231.1 | 1 | 51 |
More Information
Set Membership: |
|

Genomic Map