Assay Details
Target Gene Details
Entrez Gene ID: | 68184 |
Gene Name: | density-regulated protein |
Gene Aliases: |
1500003K04Rik |
Location: |
Chr.5:123907269-123928832 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Denr | NM_026603.4 | 8 | 1174 | NP_080879.1 |
| AK018762.1 | 8 | 1175 | BAB31392.1 | |
| AK135127.1 | 8 | 1210 | BAE22432.1 |
More Information
Set Membership: |
|

Genomic Map