Assay Details
Target Gene Details
Entrez Gene ID: | 623474 |
Gene Name: | RAD54 homolog B (S. cerevisiae) |
Gene Aliases: |
E130016E03Rik |
Location: |
Chr.4:11558920-11615808 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad54b | NM_001039556.3 | 10 | 1706 | NP_001034645.1 |
| BC057604.1 | 10 | 1654 | AAH57604.1 |

Genomic Map