Assay Details
Target Gene Details
Entrez Gene ID: | 60599 |
Gene Name: | transformation related protein 53 inducible nuclear protein 1 |
Gene Aliases: |
2700057G22Rik, SIP, SIP18, SIP27, Stinp, Teap |
Location: |
Chr.4:11156441-11174377 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53inp1 | NM_001199105.1 | 2 | 282 | NP_001186034.1 |
| NM_021897.3 | 2 | 282 | NP_068697.1 | |
| AJ131776.1 | 2 | 283 | CAB66138.1 | |
| AK054214.1 | 2 | 280 | ||
| AK142321.1 | 2 | 278 | BAE25030.1 | |
| AY034611.1 | 2 | 283 | AAK60419.1 | |
| AY034612.1 | 2 | 283 | AAK60420.1 | |
| BC132124.1 | 1 | 184 | ||
| BC138711.1 | 1 | 184 |

Genomic Map