Assay Details
Target Gene Details
Entrez Gene ID: | 14371 |
Gene Name: | frizzled homolog 9 (Drosophila) |
Gene Aliases: |
mfz9 |
Location: |
Chr.5:135248938-135251047 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fzd9 | NM_010246.1 | 1 | 1408 | NP_034376.1 |
| AF033585.2 | 1 | 1263 | AAB87503.2 | |
| AK021164.1 | 1 | 1593 | BAB32311.1 | |
| Y17709.1 | 1 | 961 | CAB44237.1 |
More Information
Set Membership: |
|

Genomic Map