Assay Details
Target Gene Details
Entrez Gene ID: | 12640 |
Gene Name: | glycoprotein hormones, alpha subunit |
Gene Aliases: |
CG-alpha, FSHA, GPHA1, GPHalpha, HCG, LHA, Tsha, aGSU, alpha-GSU, alphaGSU, alphaSU |
Location: |
Chr.4:34893779-34907374 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cga | NM_009889.2 | 4 | 481 | NP_034019.1 |
| AF307151.1 | 3 | 395 | AAG30279.1 | |
| BC087926.1 | 4 | 479 | AAH87926.1 | |
| J00643.1 | 4 | 449 | AAA96700.1 |
More Information
Set Membership: |
|

Genomic Map